Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetophenone, 99%, Spectrum™

Click to view available options
Quantity:
500 mL
200 L
Packaging:
Amber Glass Bottle
Drum
98-86-2
98-86-2
Packaging Drum
Quantity 200 L
Appearance Clear Liquid
CAS Max % 1
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.