Alfa Aesar™ Acetophenone oxime, 98%

Manufacturer: Alfa Aesar™ A1180430

Quantity 250g

 View more versions of this product

Catalog No. AAA1180430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 613-91-2
    Molecular Formula C8H9NO
    Formula Weight 135.16
    MDL Number MFCD00013931
    Beilstein 1562059
    Synonym Methyl phenyl ketoxime
    Chemical Name or Material Acetophenone oxime

    Specifications

    Formula Weight 135.16
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C8H9NO
    RTECS Number AM9654000
    CAS 613-91-2
    Health Hazard 1 Warning
    Boiling Point 118° to 120°C (20mmHg)
    TSCA TSCA
    MDL Number MFCD00013931
    Beilstein 1562059
    Synonym Methyl phenyl ketoxime
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Melting Point 58° to 60°C
    Quantity 250g
    Recommended Storage −30°C to −10°C
    Chemical Name or Material Acetophenone oxime
    Assay 98%

    Description

    Applications
    Used as a ligand in transition-metal complex catalyst chemistry. Oxime acts as an antioxidant, radical scavenger which find applications in textile, plastic, paint, detergent, and rubber industry.

    Solubility
    Slightly soluble in water. Soluble in ethyl alcohol almost transparency.

    Notes
    Air and Moisture Sensitive. Store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    GHS H Statement
    H302
    Harmful if swallowed.

    −30°C to −10°C

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit