Alfa Aesar™ Acetophenone p-toluenesulfonylhydrazone, 98%

Manufacturer: Alfa Aesar™ H5347414

Quantity 25g

 View more versions of this product

Catalog No. AAH5347414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4545-21-5
    Molecular Formula C15H16N2O2S
    Formula Weight 288.36
    MDL Number MFCD01140191
    Synonym Acetophenone tosylhydrazone; p-Toluenesulfonic acid (alpha-methylbenzylidene)hydrazide
    Chemical Name or Material Acetophenone p-toluenesulfonylhydrazone

    Specifications

    Formula Weight 288.36
    Synonym Acetophenone tosylhydrazone; p-Toluenesulfonic acid (alpha-methylbenzylidene)hydrazide
    Molecular Formula C15H16N2O2S
    MDL Number MFCD01140191
    CAS 4545-21-5
    Melting Point 144° to 149°C
    Quantity 25g
    Chemical Name or Material Acetophenone p-toluenesulfonylhydrazone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit