Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetoxime Benzoate 98.0+%, TCI America™

Click to view available options
Quantity:
25g
942-89-2
C10H11NO2
177.203
MFCD00191697
CTIVZNURROIUHT-UHFFFAOYSA-N
O-Benzoylacetoxime, O-Benzoyloxime 2-Propanone
569855
(propan-2-ylideneamino) benzoate
CC(=NOC(=O)C1=CC=CC=C1)C
942-89-2
C10H11NO2
177.203
MFCD00191697
CTIVZNURROIUHT-UHFFFAOYSA-N
O-Benzoylacetoxime, O-Benzoyloxime 2-Propanone
569855
(propan-2-ylideneamino) benzoate
CC(=NOC(=O)C1=CC=CC=C1)C
Boiling Point 120°C
Melting Point 42°C
Color White-Yellow
Quantity 25g
Formula Weight 177.20
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material Acetoxime Benzoate