Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetoxyacetone 95%, ACROS Organics™

Click to view available options
Quantity:
10mL
Packaging:
Glass bottle
592-20-1
C5H8O3
116.116
MFCD00042848
DBERHVIZRVGDFO-UHFFFAOYSA-N
acetoxyacetone,acetonyl acetate,acetol acetate,2-propanone, 1-acetyloxy,acetoxypropanone,o-acetylacetol,1-acetoxyacetone,acetylmethyl acetate,acetoxy-2-propanone,1-acetoxy-2-propanone
11593
2-oxopropyl acetate
CC(=O)COC(=O)C
592-20-1
C5H8O3
116.116
MFCD00042848
DBERHVIZRVGDFO-UHFFFAOYSA-N
acetoxyacetone,acetonyl acetate,acetol acetate,2-propanone, 1-acetyloxy,acetoxypropanone,o-acetylacetol,1-acetoxyacetone,acetylmethyl acetate,acetoxy-2-propanone,1-acetoxy-2-propanone
11593
2-oxopropyl acetate
CC(=O)COC(=O)C
Quantity 10mL
Refractive Index 1.4140 to 1.4160 (20°C, 589nm)
Packaging Glass bottle
Color Amber to Yellow
Infrared Spectrum Authentic
Density 1.0750g/mL
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula CH3C(O)OCH2C(O)CH3
Formula Weight 116.12
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material Acetoxyacetone
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning