Learn More
Acetoxymethylpolystyrene resin, 1% crossl. with DVB,0.5-0.8mmol/g,100-200 mesh, Acros Organics
Manufacturer:  Acros Organics 349480050
 View more versions of this product
Catalog No. AC349480050
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
for solid support synthesis
Specifications
Specifications
crosslinking: 1% DVB
1% Crosslinked with DVB, 0.5-0.8 mmol/g
100 to 200 mesh
White
Beads
Acetoxymethylpolystyrene resin
0.5 to 0.8mmol/g
5g
MFCD02685746