Learn More
Alfa Aesar™ Acetyl Angiotensinogen (1-14), Porcine
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66071MCR
 View more versions of this product
Catalog No. AAJ66071MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1801.08
Porcine
Solid
RUO
Acetyl Angiotensinogen (1-14), porcine
HPLC
1mg
Ac-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Leu-Val-Tyr-Ser
-30° to -10°C