Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl bromide, 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
506-96-7
C2H3BrO
122.95
MFCD00000114
FXXACINHVKSMDR-UHFFFAOYSA-N
ethanoyl bromide,acetylbromide,acetic acid, bromide,ch3cobr,unii-o18v5xyo0g,hsdb 663,o18v5xyo0g,bromideacetyl,acbr,acetyl bromide
10482
acetyl bromide
CC(=O)Br
506-96-7
C2H3BrO
122.95
MFCD00000114
FXXACINHVKSMDR-UHFFFAOYSA-N
ethanoyl bromide,acetylbromide,acetic acid, bromide,ch3cobr,unii-o18v5xyo0g,hsdb 663,o18v5xyo0g,bromideacetyl,acbr,acetyl bromide
10482
acetyl bromide
CC(=O)Br
Density 1.6600g/mL
Flash Point 75°C
Boiling Point 75°C to 77°C
Melting Point -96°C
Specific Gravity 1.66
Refractive Index 1.4485 to 1.4505
Packaging Glass bottle
Quantity 100mL
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula CH3COBr
Beilstein 02, 174
Fieser 06,9; 13,208
Merck Index 15, 77
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohol and acetone,soluble in ether,chloroform and benzene
Formula Weight 122.95
Physical Form Fuming Liquid
Percent Purity 98%
Chemical Name or Material Acetyl bromide, 98%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 208-061-7

RTECSNumber : AO5955000

TSCA : TSCA

Recommended Storage : May darken during storage