Alfa Aesar™ Acetyl chloride, 1M soln. in dichloromethane

Manufacturer: Alfa Aesar™ H32616AP

Quantity 500mL

 View more versions of this product

Catalog No. AAH32616AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75-36-5
    Molecular Formula CH3COCl
    Formula Weight 78.5
    MDL Number MFCD00000719
    UN Number UN1593
    Chemical Name or Material Acetyl chloride

    Specifications

    Formula Weight 78.5
    TSCA TSCA
    Molecular Formula CH3COCl
    MDL Number MFCD00000719
    CAS 75-36-5
    UN Number UN1593
    Concentration 1M soln. in dichloromethane
    Quantity 500mL
    Health Hazard 2 GHS H Statement
    H351-H302-H315-H319
    Suspected of causing cancer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P281-P305+P351+P338-P362-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DICHLOROMETHANE
    Chemical Name or Material Acetyl chloride

    GHS H Statement
    H351-H302-H315-H319
    Suspected of causing cancer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P281-P305+P351+P338-P362-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit