Learn More
Alfa Aesar™ Acetyl chloride, 1M soln. in dichloromethane
Manufacturer:  Alfa Aesar™ H32616AE
Quantity 100mL
 View more versions of this product
Catalog No. AAH32616AE
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
75-36-5
78.5
UN1593
CH3COCl
MFCD00000719
Acetyl chloride
Specifications
75-36-5
78.5
UN1593
100mL
CH3COCl
MFCD00000719
1M soln. in dichloromethane
Acetyl chloride
Safety and Handling
SDS & Safety Documents
GHS H Statement
H351-H302-H315-H319
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280-P281-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DICHLOROMETHANE
TSCA : TSCA