Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl chloride, 1M solution in dichloromethane, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
75-36-5,75-09-2
C2H3ClO
78.495
MFCD00000719
WETWJCDKMRHUPV-UHFFFAOYSA-N
ethanoyl chloride,acetic chloride,acetylchloride,acetic acid chloride,ch3cocl,rcra waste number u006,acetic acid, chloride,unii-qd15rno45k,ccris 4568,hsdb 662
6367
CHEBI:37580
acetyl chloride
CC(=O)Cl
75-36-5,75-09-2
C2H3ClO
78.495
MFCD00000719
WETWJCDKMRHUPV-UHFFFAOYSA-N
ethanoyl chloride,acetic chloride,acetylchloride,acetic acid chloride,ch3cocl,rcra waste number u006,acetic acid, chloride,unii-qd15rno45k,ccris 4568,hsdb 662
6367
CHEBI:37580
acetyl chloride
CC(=O)Cl
CAS Min % 5.44,93.36
CAS Max % 6.64,94.57
Linear Formula CH3COCl
Concentration 0.9 to 1.1M
Merck Index 15, 80
Density 1.3000g/mL
Formula Weight 78.5
Physical Form Liquid
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: reacts with water.
Specific Gravity 1.3
Color Colorless
Quantity 800mL
Chemical Name or Material Acetyl chloride, 1M solution in dichloromethane
Show More Show Less

GHS H Statement
Suspected of causing cancer if inhaled. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Reacts violently with water. Corrosive to the respiratory tract.

GHS P Statement
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Danger