Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Acetyl-CoA Synthetase, Mouse, Unlabeled, Clone: 6C1.2,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABS1720
Description
Acetyl-coenzyme A synthetase, cytoplasmic (EC 6.2.1.1; UniProt Q9NR19; also known as Acetate-CoA ligase, Acetyl-CoA synthetase, AceCS, ACS, Acyl-activating enzyme, Acyl-CoA synthetase short-chain family member 2) is encoded by the ACSS2 (also known as ACAS2) gene (Gene ID 55902) in human. ACS plays a vital role in prokaryotic and eukaryotic cells by activating acetate to form acetyl CoA that serves as a source of two carbon metabolite for lipid synthesis and energy producing reactions. It is also reported to play a role in gene expression. In yeast cells it delivers acetyl CoA for histone acetylation. ACS contributes to cancer cell growth under low-oxygen and lipid-depleted conditions and its expression is upregulated under metabolically stress. Silencing ACSS2 gene is shown to reduce the growth of tumor xenografts. Ref.: Starai, VJ., and Escalante-Semerena, JC. (2004). Cell. Mol. Life Sci. 61, 2020 2030. Schug, ZT et al. (2015). Cancer Cell. 27:57-71.
Specifications
Acetyl-CoA Synthetase | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide. | |
Q9NR19 | |
GST-tagged recombinant human acetyl-CoA synthetase N-terminal fragment. | |
100 μg | |
Signaling | |
NP_061147 | |
Human | |
Purified |
Western Blot | |
6C1.2 | |
Unconjugated | |
ACSS2, ACAS2 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Immunogen sequence is present in both spliced isoforms of human acetyl-CoA synthetase reported by UniProt (Q9NR19). | |
Stable for 1 year at 2°-8°C from date of receipt. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction