Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl Histone H3 (Lys9 and 14) clone 5H20 Rabbit anti-Human,Mouse,Rat, KHL, Clone: 5H20, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen Acetyl Histone H3 (Lys9 and 14) clone 5H20
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibition Assays, Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB065992MI
View Documents
MilliporeSigma
ZRB0659925UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB065994MI
View Documents
MilliporeSigma
ZRB065994X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Acetyl Histone H3 (Lys9 and 14) clone 5H20 Monoclonal antibody specifically detects Acetyl Histone H3 (Lys9 and 14) clone 5H20 in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Peptide Inhibition Assay, Western Blotting.
Specifications

Specifications

Acetyl Histone H3 (Lys9 and 14) clone 5H20
KHL
Rabbit
H3K9;14Ac;Histone H3 Ac Lysine 9;14;Histone H3.1t;H3/t;H3t;H3/g
KLH-conjugated linear peptide corresponding to the first 21 amino acids (without methionine) surrounding acetylated lysines 9 and 14 of human Histone H3.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibition Assays, Western Blot
Purified
Human, Mouse, Rat
HIST3H3, H3FT
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit