Learn More
Alfa Aesar™ Acetyl-Myelin Basic Protein (4-14)Peptide
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66946EXD
 View more versions of this product
Catalog No. AAJ66946EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1432.70
0.5mg
N-Acetyl-Gln-Lys-Arg-Pro-Ser-Gln-Arg-Ser-Lys-Tyr-Leu
-30° to -10°C
Acetyl-Myelin Basic Protein (4-14) Peptide
HPLC
Solid
RUO
Ac-MBP (4-14) Peptide