Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Acetylacetone, ≥99.6% (GC), MilliporeSigma™ Supelco™

Acetylacetone is a β-diketone, which contains two carbonyl groups and one active methylene group.

Specifications

CAS 123-54-6
Percent Purity ≥99.6% (GC)
Density 0.975 g/mL (at 25°C)
Grade Analytical Standard
Linear Formula CH3 COCH2 COCH3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-102-0549
View Documents
MilliporeSigma Supelco
055811MLF
1 mL
Each for $27.35
N/A
11-101-9095
View Documents
MilliporeSigma Supelco
055815MLF
5 mL
Each for $105.90
N/A
Description

Description

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications

Specifications

123-54-6
0.975 g/mL (at 25°C)
CH3 COCH2 COCH3
≥99.6% (GC)
Analytical Standard
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit