Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Affymetrix/USB™ Acetylated Bovine Albumin

Bovine Serum Albumin is used by molecular biologists to stabilize labile enzymes such as restriction endonucleases, polymerases, and ligases.

Manufacturer:  Affymetrix/USB™ J10848MB

 View more versions of this product

Catalog No. AAJ10848MB

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Source: Bovine blood
  • Form: Clear, colorless solution in deionized water
  • Assay: Single band by SDS-polyacrylamide gel electrophoresis
  • Protein concentration: 25 - 50mg/mL
  • DNase (endo), DNase (exo), RNase, Protease: Passes test
  • No inhibition by albumin on digestion of lambda DNA
  • Solubility: Soluble in water
  • Storage: -20°C
  • CAS: 9048-46-8
  • Packed under aseptic, nuclease-free conditions
Specifications

Specifications

25 - 50mg/mL
Solution
Bovine blood
Acetylated Bovine Albumin
Stabilize labile enzymes such as restriction endonucleases, polymerases, and ligases
25mg
Plastic bottle
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit