Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylated Lysine Mouse anti-Mouse, APC, Clone:7F8, Abnova™

Mouse monoclonal antibody raised against Acetylated Lysine.

Manufacturer:  Abnova Corporation MAB16960

Catalog No. 89-333-690


Add to cart

Specifications

Specifications

Acetylated Lysine
7F8
ELISA
Immunocytochemistry
Immunofluorescence
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100)
Immunoprecipitation
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Liquid
Acetylated KLH.
100μg
Antibody
Mouse
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
APC
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Mouse
IgG1
Store at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit