Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(±)-Acetylcarnitine chloride, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
Weak cholinergic agonist. Acylcarnitines are important intermediates in lipid metabolism.
Specifications
Quantity | 100 mg |
Formula Weight | 239.7 |
Percent Purity | >98% |
Chemical Name or Material | (±)-Acetylcarnitine chloride |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title