Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetylcholine Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10064656AF750
Description
Acetylcholine Polyclonal antibody specifically detects Acetylcholine in All samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)Specifications
Acetylcholine | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
All species | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Alexa Fluor 750 | |
[2 (acetyloxy)ethyl]trimethylammonium, ACh | |
Synthetic acetylcholine conjugated to Keyhole limpet Haemocyanin. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction