Alfa Aesar™ Acetylcholine iodide, 98%

Manufacturer: Alfa Aesar™ A1169422

Quantity 100g

 View more versions of this product

Catalog No. AAA1169422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2260-50-6
    Molecular Formula CH3CO2CH2CH2N(CH3)3I
    Formula Weight 273.11
    MDL Number MFCD00011815
    Beilstein 3571339
    Synonym (2-Acetoxyethyl)trimethylammonium iodide; 2-Acetoxy-N,N,N-trimethylethanium iodide
    Chemical Name or Material Acetylcholine iodide

    Specifications

    Formula Weight 273.11
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011815
    Melting Point 161° to 165°C
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Recommended Storage 2°C to 8°C
    TSCA TSCA
    CAS 2260-50-6
    Synonym (2-Acetoxyethyl)trimethylammonium iodide; 2-Acetoxy-N,N,N-trimethylethanium iodide
    Health Hazard 1 Warning
    Quantity 100g
    RTECS Number KH3300000
    Molecular Formula CH3CO2CH2CH2N(CH3)3I
    Beilstein 3571339
    Chemical Name or Material Acetylcholine iodide
    Assay 98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit