Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Acetylcholinesterase/ACHE Mouse anti-Human, Bovine, Janelia Fluor 646, Clone: 400000000000, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP105146JF646
Catalog No. NBP15146646
Description
Description
Acetylcholinesterase/ACHE Monoclonal antibody specifically detects Acetylcholinesterase/ACHE in Human, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
Specifications
Acetylcholinesterase/ACHE
400000000000
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
ACHE
C-terminal 10 residues of brain acetylcholinesterase (human and bovine), absent from the erythrocyte enzyme.
Protein A or G purified
Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary
43
Bovine, Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Specific for brain AChE and does not recognize AChE from erythrocytes. The can thus distinguish between mammalian brain AChE and erythrocyte AChE. Weak cross-reactivity with Torpedo marmorata AChE but none with AChE from electric eel or human BtChE.