Learn More
Selleck Chemical LLCSupplier Diversity Partner Acetylcysteine (N-acetylcysteine) 50mg 616-91-1 NAC  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S162350MG
Acetylcysteine (N-acetyl-l-cysteine, NAC) is a ROS(reactive oxygen species) inhibitor that antagonizes the activity of proteasome inhibitors. It is also a tumor necrosis factor production inhibitor. Acetylcysteine(N-acetyl-l-cysteine) suppresses TNF-induced NF-κB activation through inhibition of IκB kinases. Acetylcysteine(N-acetyl-l-cysteine) induces apoptosis via the mitochondria-dependent pathway. Acetylcysteine(N-acetyl-l-cysteine) inhibits ferroptosis and virus replication.Solutions of Acetylcysteine are best fresh-prepared.
Catalog No. 50-714-3