Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylcysteine, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
616-91-1
C5H9NO3S
163.19
PWKSKIMOESPYIA-UHFFFAOYNA-N
2-acetamido-3-sulfanylpropanoic acid
CC(=O)NC(CS)C(O)=O
616-91-1
C5H9NO3S
163.19
PWKSKIMOESPYIA-UHFFFAOYNA-N
2-acetamido-3-sulfanylpropanoic acid
CC(=O)NC(CS)C(O)=O

AC126, 616-91-1, C5H9NO3S

Spectrum™ Acetylcysteine, USP is used as a nutritional antioxidant, mucolytic therapy agent, nephroprotective agent and acetaminophen overdose antitoxin. All Spectrum Chemical USP products, including all USP Acetylcysteine products, are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Ignition Residue 0.5%
Loss on Drying 1%
Packaging Fiber Drum
Specific Rotation +21° to +27°
Percent Purity 98 to 102%
Quantity 25 kg