Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylurea 99.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
591-07-1
C3H6N2O2
102.093
MFCD00038140
GKRZNOGGALENQJ-UHFFFAOYSA-N
acetylurea,acetamide, n-aminocarbonyl,n-acetylurea,acetylcarbamide,monoacetylurea,1-acetylurea,acetic acid, ureide,n-aminocarbonyl acetamide,urea, acetyl,unii-8kru31i919
68956
N-carbamoylacetamide
CC(=O)NC(=O)N
591-07-1
C3H6N2O2
102.093
MFCD00038140
GKRZNOGGALENQJ-UHFFFAOYSA-N
acetylurea,acetamide, n-aminocarbonyl,n-acetylurea,acetylcarbamide,monoacetylurea,1-acetylurea,acetic acid, ureide,n-aminocarbonyl acetamide,urea, acetyl,unii-8kru31i919
68956
N-carbamoylacetamide
CC(=O)NC(=O)N
Quantity 500g
Melting Point 215°C
Color White
Formula Weight 102.09
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.0% (N)
Chemical Name or Material Acetylurea

RTECSNumber : AB4331500

TSCA : Yes