Learn More
Medchemexpress LLC AChE-IN-26 | 2977170-10-6 | C21H21BrN2O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1513705MG
Encompass_Preferred
AChE-IN-26 (compound 4a) is an AChE (acetylcholinesterase) inhibitor with an IC50 value of 0.42 μM. It can be used for the research of Alzheimer's disease.
  • AChE (acetylcholinesterase) inhibitor
  • IC50 value of 0.42 μM
  • Suitable for Alzheimer's disease research
Catalog No. 50-003-72215