Learn More
Pyramex™ Achieva™ Safety Eyewear
Lightweight, frameless protection is ideal for all-day wear
$6.46 - $7.94
Specifications
Lens Material Polycarbonate
Description 8 base curve lens , wrap around design with side protection
Certifications/Compliance ANSI Z87.1
View More Specs
Products 4
Catalog Number Mfr. No. Temple Color Lens Tint Lens Type Price Quantity & Availability  
Catalog Number Mfr. No. Temple Color Lens Tint Lens Type Price Quantity & Availability  
19-155-933
View Documents
Pyramex™
SG6510S
Clear with Gray Tips Clear Anti-UV
Each for $6.46
Case of 12 EA for $73.90
Add to cart
19-155-934
View Documents
Pyramex™
SG6510ST
Clear with Gray Tips Clear Anti-fog, Anti-UV
Each for $7.94
Case of 12 EA for $90.80
Add to cart
19-155-935
View Documents
Pyramex™
SG6520S
Gray with Gray Tips Gray Anti-UV
Each for $7.18
Case of 12 EA for $82.10
Add to cart
19-155-975
View Documents
Pyramex™
SP6510S
Clear with Pink Tips Clear Anti-UV
Each for $6.46
Case of 12 EA for $73.90
Add to cart
Description
Description
  • Brow guard deflects airborne particles
  • Built-in rubber nose piece provides all day comfort
  • Unique rubber temple tips twist around the temples for a stylish and secure fit
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Specifications
    Polycarbonate
    8 base curve lens , wrap around design with side protection
    ANSI Z87.1