Learn More
EVOS™ EVOS™ 100X Oil Objective, achromat, coverslip-corrected
For general applications and image with different magnification
Supplier: EVOS™ AMPFOP100
Description
- Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
- Offers optical performance from visible light to near infrared light
- Meets needs across spectrum of magnifications and optical specifications
Specifications
Objective Lens | |
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System | |
0.17 mm | |
100X | |
1 Ea. |
EVOS | |
Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates | |
Achromat | |
1.25 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.