Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acid Alcohol, Orth's Decolorizing Fluid, 3% HCl in 70% Alcohol, Ricca Chemical
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
Specifications
Physical Form | Liquid |
CAS Min % | 26.0, 66.0, 3.0, 2.9 |
CAS Max % | 28.0, 67.0, 4.0, 3.1 |
Packaging | Natural Poly Bottle |
Color | Colorless |
Quantity | 1 L |
Concentration or Composition (by Analyte or Components) | 3% HCl |
Chemical Name or Material | Acid Alcohol, Orth's Decolorizing Fluid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction