Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Blue 20, TCI America™

Click to view available options
Quantity:
25g
8004-99-7
MFCD00131375
Induline (Water soluble)
8004-99-7
MFCD00131375
Induline (Water soluble)
Physical Form Crystal-Powder at 20°C
Color Black
Quantity 25g
Chemical Name or Material Acid Blue 20