Learn More
Getinge USA Acid Detergent 4x1 gallon  DFS Item
Manufacturer:  Getinge USA 61301605278
Strong dissolving action against minerals and scales. Cleans instruments, utensils, and glassware. Removes residues from washer-disinfectors and other similar washers.
Catalog No. NC0578427