Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Green 41, pure, Acros Organics

Click to view available options
Quantity:
25g
4430-16-4
C28H20N2Na2O10S2
654.572
MFCD00011715
LERIXBKRKBHLHG-UHFFFAOYSA-L
C.I. 62560
78155
disodium;2-[[5,8-dihydroxy-4-(4-methyl-2-sulfonatoanilino)-9,10-dioxoanthracen-1-yl]amino]-5-methylbenzenesulfonate
CC1=CC(=C(C=C1)NC2=C3C(=C(C=C2)NC4=C(C=C(C=C4)C)S(=O)(=O)[O-])C(=O)C5=C(C=CC(=C5C3=O)O)O)S(=O)(=O)[O-].[Na+].[Na+]
4430-16-4
C28H20N2Na2O10S2
654.572
MFCD00011715
LERIXBKRKBHLHG-UHFFFAOYSA-L
C.I. 62560
78155
disodium;2-[[5,8-dihydroxy-4-(4-methyl-2-sulfonatoanilino)-9,10-dioxoanthracen-1-yl]amino]-5-methylbenzenesulfonate
CC1=CC(=C(C=C1)NC2=C3C(=C(C=C2)NC4=C(C=C(C=C4)C)S(=O)(=O)[O-])C(=O)C5=C(C=CC(=C5C3=O)O)O)S(=O)(=O)[O-].[Na+].[Na+]
Infrared Spectrum Authentic
Formula Weight 654.57
Physical Form Crystalline Powder
Grade Extra Pure
UV Spectrometry Complies
Color Green
Quantity 25g
Chemical Name or Material Acid green 41