Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acid Value Solution, Reagents
Acid Value Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$76.00 - $121.00
Specifications
Chemical Name or Material | Acid Value Solution |
---|---|
Color | Colorless to Light Pink |
CAS | 1330-20-7, 67-63-0, 67-63-0 |
Synonym | Xylenes/Isopropanol |
Grade | Reagent |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-335-988
|
Reagents Holdings Llc
CA0400002.5C |
2.5 L |
Each for $76.00
|
|
|||||
01-335-987
|
Reagents Holdings Llc
CA0400004C |
4 L |
Each for $121.00
|
|
|||||
Specifications
Acid Value Solution | |
1330-20-7, 67-63-0, 67-63-0 | |
Reagent | |
0.8413 | |
Liquid |
Colorless to Light Pink | |
Xylenes/Isopropanol | |
Glass Bottle | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title