Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Yellow 11, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
6359-82-6
C16H14N4NaO4S
381.362
MFCD00003122
JQVFPNQVPUYAHJ-UHFFFAOYSA-N
Flavazin L,C.I. 18820
131856598
4-(3-methyl-5-oxo-4-phenyldiazenyl-4H-pyrazol-1-yl)benzenesulfonic acid;sodium
CC1=NN(C(=O)C1N=NC2=CC=CC=C2)C3=CC=C(C=C3)S(=O)(=O)O.[Na]
6359-82-6
C16H14N4NaO4S
381.362
MFCD00003122
JQVFPNQVPUYAHJ-UHFFFAOYSA-N
Flavazin L,C.I. 18820
131856598
4-(3-methyl-5-oxo-4-phenyldiazenyl-4H-pyrazol-1-yl)benzenesulfonic acid;sodium
CC1=NN(C(=O)C1N=NC2=CC=CC=C2)C3=CC=C(C=C3)S(=O)(=O)O.[Na]
Beilstein 24, 337
Infrared Spectrum Authentic
Loss on Drying 7% max.
Formula Weight 380.36
Physical Form Powder
Packaging Glass bottle
Color Orange to Yellow
Quantity 25g
Chemical Name or Material Acid Yellow 11

GHS H Statement:
Causes severe skin burns and eye damage.
Risk of explosion if heated under confinement.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger