Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acinus Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$177.16 - $378.00
Specifications
Antigen | Acinus |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 -1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Description
Acinus Polyclonal specifically detects Acinus in Human samples. It is validated for Western Blot.Specifications
Acinus | |
Western Blot 1:500 -1:1000 | |
Polyclonal | |
Rabbit | |
Apoptosis, Cancer, Chromatin Research, Tumor Suppressors | |
PBS with 0.02% Sodium Azide | |
22985 | |
A peptide made to the C-terminal sequence RNRQLEREKRREHS. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Acinus, ACN, apoptotic chromatin condensation inducer 1, apoptotic chromatin condensation inducer in the nucleus, fSAP152, functional spliceosome-associated protein 152, KIAA0670DKFZp667N107 | |
ACIN1 | |
IgG | |
Affinity Purified | |
Reacts with the C-terminus of Acinus. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title