Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AcmNPV gp64 Protein Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326084H
Description
AcmNPV gp64 Protein Polyclonal antibody specifically detects AcmNPV gp64 Protein in Virus samples. It is validated for ELISASpecifications
AcmNPV gp64 Protein | |
Polyclonal | |
ELISA | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Virus | |
Purified |
ELISA | |
HRP | |
PBS | |
Rabbits immunized with purified, recombinant AcmNPV gp64 Protein (Accession#: NP_054158.1, Met1-Thr481) | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction