Learn More
Acorn™ Temp 6 RTD Thermometer
Compact, rugged meters with splash-resistant membrane keypad
 View more versions of this product
Catalog No. 13-621-853
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
±0.2°C
5.5 in.
5.5 x 2.75 x 1.25 in. (14 x 7 x 3.17cm)
3.17cm
14cm
-328° to +1562°F
16 oz.
2.75 in.
Acorn Temp 6 RTD Thermometer Kit
LCD
CE, NIST
1.25 in.
Meter, NIST certification, four 1.5V AAA battery, splash resistant membrane keypad
1°C
-200° to +850°C
450g
7cm