Learn More
Acorn™ Temp Thermocouple Meters
Rugged, multi-type thermocouple thermometers are economical, compact, and dependable
 View more versions of this product
Catalog No. 13-621-841
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Requires : Use with Digi-Sense™ thermocouple probes
Specifications Description & Specifications
Specifications
0.5°C
5.5 in.
5.5 x 2.7 x 1.3 in. (14 x 6.85 x 3.3cm)
3.3cm
14cm
1°C
450g
6.85cm
LCD
CE, NIST
1.3 in.
Meter, general purpose probe, NIST Traceable calibration report, Rubber armor and four 1.5V AAA batteries, for more than 200 hours of continuous use
Thermocouple probe Type-J, K or T
16 oz.
2.7 in.
Accessories