Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACOT11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565282
Description
Immunogen sequence: FLLLSDLRQRP EWDKHYRSVE LVQQVDEDDA IYHVTSPALG GHTKPQDFVI LASRRKPCDN GDPYVIALRS VTLPTHRETP EYRRGE Highest antigen sequence identity to the following orthologs - mouse 92%, rat 89%.
This gene encodes a member of the acyl-CoA thioesterase family which catalyse the conversion of activated fatty acids to the corresponding non-esterified fatty acid and coenzyme A. Expression of a mouse homolog in brown adipose tissue is induced by low temperatures and repressed by warm temperatures. Higher levels of expression of the mouse homolog has been found in obesity-resistant mice compared with obesity-prone mice, suggesting a role of acyl-CoA thioesterase 11 in obesity. Alternative splicing results in transcript variants.
Specifications
ACOT11 | |
Polyclonal | |
Unconjugated | |
ACOT11 | |
1110020M10Rik; 2010309H15Rik; ACOT11; Acyl-CoA thioester hydrolase 11; acyl-CoA thioesterase 11; Acyl-coenzyme A thioesterase 11; adipose-associated thioesterase; AW060409; Bfit; BFIT1; Brown fat-inducible thioesterase; KIAA0707; mKIAA0707; Palmitoyl-coenzyme A thioesterase; STARD14; StAR-related lipid transfer (START) domain containing 14; START domain containing 14; Thea; Them1; thioeste; thioesterase superfamily member 1; thioesterase, adipose associated | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26027 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WXI4 | |
ACOT11 | |
Recombinant Human ACOT11. Recombinant protein control fragment (Product #RP-104727). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction