Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACOT12 Mouse anti-Human, Clone: OTI1B9, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502462

Catalog No. 50-167-3514


Add to cart

Description

Description

Hydrolyzes acetyl-CoA to acetate and CoA.
Specifications

Specifications

ACOT12
Monoclonal
0.91 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8WYK0
CACH-1, Cach, STARD15, THEAL
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
134526
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI1B9
Unconjugated
ACOT12
Liquid
ACOT12
Full length human recombit protein of human ACOT12 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit