Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACOT12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549968
Description
The antibody detects endogenous levels of total ACOT12 protein.
Hydrolyzes acetyl-CoA to acetate and CoA.
Specifications
ACOT12 | |
Polyclonal | |
Unconjugated | |
ACOT12 | |
1300004O04Rik; 4930449F15Rik; Acetyl-coenzyme A thioesterase; Acot12; Acyl-CoA thioester hydrolase 12; acyl-CoA thioesterase 12; Acyl-coenzyme A thioesterase 12; AV027244; Cach; CACH1; CACH-1; Cytoplasmic acetyl-CoA hydrolase 1; cytosolic acetyl-CoA hydrolase; hCACH-1; mCACH-1; rACH; rCACH-1; STARD15; StAR-related lipid transfer (START) domain containing 15; START domain-containing protein 15; THEAL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
134526 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8WYK0 | |
ACOT12 | |
Synthesized peptide derived from internal of human ACOT12 (aa 281-330). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction