Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACOT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101403
Description
Antibody detects endogenous levels of total ACOT2.
Acyl-CoA thioesterases (ACOTs) are a group of enzymes that catalyze the hydrolysis of acyl-CoA to form coenzyme A (CoA) and a free fatty acid. Through their catalytic activity, ACOTs are able to regulate the level of fatty acids and acyl-CoAs within the cell. ACOT1 (acyl-CoA thioesterase 1, also known as CTE1) and ACOT2 (acyl-CoA thioesterase 2, also known as PTE2) are members of the ACOT family and exhibit different cellular localization, with ACOT1 existing as a monomer in the cytoplasm and ACOT2 localized primarily to mitochondria. Characteristic of most ACOT proteins, ACOT1 and ACOT2 catalyze the conversion of Palmitoyl-CoA and water to free CoA and palmitate, a reaction that is important for the regulation of intercellular fatty acid levels. ACOT2 is expressed as multiple alternatively spliced isoforms and, like ACOT1, is encoded by a gene which maps to human chromosome 14.
Specifications
ACOT2 | |
Polyclonal | |
Unconjugated | |
ACOT2 | |
AA571646; Acot2; Acyl coenzyme A thioester hydrolase; acyl-CoA thioesterase 2; Acyl-coenzyme A thioester hydrolase 2a; acyl-coenzyme A thioesterase 2, mitochondrial; ARTISt/p43; CTE Ia; CTE1A; CTE-Ia; long-chain acyl-CoA thioesterase 2; mitochondrial acyl-CoA thioesterase 1; Mte1; MTE-I; peroxisomal long-chain acyl-coA thioesterase 2; PTE2; PTE2A; Very-long-chain acyl-CoA thioesterase; ZAP128 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10965 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P49753 | |
ACOT2 | |
A synthesized peptide derived from human ACOT2(Accession P49753), corresponding to amino acid residues D177-G227. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction