Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACOT7 Antibody (1D5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222569
Description
ACOT7 Monoclonal specifically detects ACOT7 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
ACOT7 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Flow Cytometry, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
ACH1, acyl-CoA thioesterase 7CTE-IIa, BACHEC 3.1.2.2, brain acyl CoA hydrolase, Brain acyl-CoA hydrolase, CTE-IILACH, cytosolic acyl coenzyme A thioester hydrolase, hBACH, long chain, Long chain acyl-CoA thioester hydrolase | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Anti-human ACOT7 mAb, clone 1D5, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human ACOT7 protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
1D5 | |
Unconjugated | |
NP_009205 | |
ACOT7 | |
Recombinant human ACOT7 (1-370aa) purified from E. coli | |
0.1 mL | |
Neuroscience, Signal Transduction | |
11332 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction