Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACOT9 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549048
Description
The protein encoded by this gene is a mitochondrial acyl-CoA thioesterase of unknown function. Two transcript variants encoding different isoforms have been found for this gene.Specifications
ACOT9 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9Y305, Q9R0X4 | |
ACOT9 | |
KLH conjugated synthetic peptide between 213-240 amino acids from the Central region of human ACOT9. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ACOT9 | |
Acyl-coenzyme A thioesterase 9, mitochondrial, Acyl-CoA thioesterase 9, 312-, Acyl-CoA thioester hydrolase 9, ACOT9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23597, 56360 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction