Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACOXL Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 233661AP

Catalog No. 50-173-4060


Add to cart

Description

Description

ACOXL gene ontology annotations related to this gene include fatty acid beta-oxidation.
Specifications

Specifications

ACOXL
0.45 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NUZ1
ACOXL, Acyl CoA oxidase like protein, acyl Coenzyme A oxidase like
Rabbit
IgG
150 μL
-20°C
Primary
55289
Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
ACOXL
Liquid
ACOXL
ACOXL Fusion Protein Ag18625
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit