Learn More
ACP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 152361AP
Catalog No. 50-173-0085
Add to cart
Description
Description
This gene encodes the beta subunit of lysosomal acid phosphatase (LAP). LAP is chemically and genetically distinct from red cell acid phosphatase. The encoded protein belongs to a family of distinct isoenzymes which hydrolyze orthophosphoric monoesters to alcohol and phosphate. Mutations in this gene or in the related alpha subunit gene cause acid phosphatase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
Specifications
Specifications
ACP2
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P11117, P20611, P24638
acid phosphatase 2, lysosomal, ACP2, LAP, Lysosomal acid phosphatase
Rabbit
IgG
150 μL
-20°C
Primary
11432, 24162, 53
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
ACP2
Liquid
ACP2
ACP2 Fusion Protein Ag7356
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat