Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACP5 Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: ACP5/2336R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00602

Catalog No. 89-339-581


Add to cart

Description

Description

ACP5 Monoclonal antibody specifically detects ACP5 in Human, Mouse, Rat samples. It is validated for IHC-P.
Specifications

Specifications

ACP5
ACP5/2336R
acid phosphatase 5, tartrate resistant
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin)
Unconjugated
ACP5
MGC117378, TRAP
Recombinant protein corresponding to full-length human ACP5
100 μg
Primary
54
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit