Learn More
Abnova™ ACP6 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Supplier:  Abnova™ H00051205T02
Catalog No. 89-932-373
Add to Cart
Description
Description
Specifications
Specifications
Western Blot
ACP6 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 48.9kDa
Transient overexpression cell lysate was tested with anti-ACP6 antibody (Mfr. No. H00051205-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
293T
NM_016361.2
pCMV-ACP6 full-length
100 μL
Human
Product Suggestions
Product Suggestions