ACPA (in Tocrisolve 100), Tocris

Potent, selective CB1 agonist (in water-soluble emulsion)

Manufacturer: R&D Systems 1781/25

Quantity 25mg

 View more versions of this product

Catalog No. 17-812-5

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C23H37NO
    Formula Weight 343.55
    Synonym N-(Cyclopropyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
    Chemical Name or Material ACPA (in Tocrisolve™ 100)

    Specifications

    Formula Weight 343.55
    Quantity 25mg
    Synonym N-(Cyclopropyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
    Assay Percent Range >98%
    Molecular Formula C23H37NO
    CAS 229021-64-1
    Recommended Storage Store at +4°C
    Chemical Name or Material ACPA (in Tocrisolve™ 100)

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit