Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACPA, Tocris

Potent, selective CB1 agonist

Manufacturer:  R&D Systems 1318/5

Quantity 5mg

 View more versions of this product

Catalog No. 13-185

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

>98%
343.55
ACPA
C23H37NO
N-(Cyclopropyl)-5Z,8Z,11Z,14Z-eicosatetraenamide

Specifications

5mg
>98%
343.55
ACPA
229021-64-1
C23H37NO
N-(Cyclopropyl)-5Z,8Z,11Z,14Z-eicosatetraenamide

Safety and Handling

Recommended Storage : Store at -20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit