Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACPL2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548756
Description
Predicted to react with mouse and rat based on sequence homology.
Responsible for the 2-O-dephosphorylation of xylose in the glycosaminoglycan-protein linkage region of proteoglycans thereby regulating the amount of mature glycosaminoglycan (GAG) chains. Sulfated glycosaminoglycans (GAGs), including heparan sulfate and chondroitin sulfate, are synthesized on the so-called common GAG-protein linkage region (GlcUAbeta1-3Galbeta1-3Galbeta1- 4Xylbeta1-O-Ser) of core proteins, which is formed by the stepwise addition of monosaccharide residues by the respective specific glycosyltransferases. Xylose 2-O-dephosphorylation during completion of linkage region formation is a prerequisite for the initiation and efficient elongation of the repeating disaccharide region of GAG chains.
Specifications
ACPL2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8TE99 | |
PXYLP1 | |
KLH conjugated synthetic peptide between 306-335 amino acids from the Central region of human ACPL2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PXYLP1 | |
2-phosphoxylose phosphatase 1, 313-, Acid phosphatase-like protein 2, Xylosyl phosphatase, epididymis luminal protein 124 {ECO:0000303/Ref2, ECO:0000312/EMBL:ACJ137311}, PXYLP1 (HGNC:26303) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92370 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction